missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles

High wet strength due to the addition of chemically stable resin

Brand:  Cytiva 1091-185

Code : 69

Additional Details : Weight : 1.85000kg

 View more versions of this product

Product Code. 11352875

  • £177.00 / Pack of 1000
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

High wet strength due to the addition of chemically stable resin

  • Wet strengthened grade
  • General purpose creped filter
  • For less critical routine analysis
  • Used to assay sucrose in cane sugar

Specifications

Specifications

Grade 91 Qualitative Filter Paper Wet-Strengthened, circle, 185 mm (subdivided into 100)
Cellulose
10 μm
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
1000 Pieces (subdivided into 100)
Qualitative Filter Paper
7.28 in.
Disc
10 μm
70 sec.
Grade 91
205 μm
Ø 185 mm
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers