missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles
High wet strength due to the addition of chemically stable resin
£120.00 - £387.00
Specifications
Format | Disc |
---|---|
Material | Cellulose |
Particle Retention | 10 μm |
Pore Size | 10 μm |
Filtration Speed | 70 sec. |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
11322875
|
Cytiva
1091-110 |
110 mm | 4000 Pieces (subdivided into 100) |
£328.00
Pack of 4000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11370524
|
Cytiva
1091-165 |
165 mm | 1000 Pieces (subdivided into 100) |
£151.00
Pack of 1000 |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
11332875
|
Cytiva
1091-125 |
125 mm | 4000 Pieces (subdivided into 100) |
£387.00
Pack of 4000 |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
11380524
|
Cytiva
1091-190 |
190 mm | 1000 Pieces (subdivided into 100) |
£294.00
Pack of 1000 |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
11372875
|
Cytiva
1091-240 |
240 mm | 1000 Pieces (subdivided into 100) |
£292.00
Pack of 1000 |
Estimated Shipment: 21-11-2024
Log in to see stock. |
||||
11342875
|
Cytiva
1091-150 |
150 mm | 1000 Pieces (subdivided into 100) |
£120.00
Pack of 1000 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
11352875
|
Cytiva
1091-185 |
185 mm | 1000 Pieces (subdivided into 100) |
£177.00
Pack of 1000 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
Description
High wet strength due to the addition of chemically stable resin
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Disc | |
10 μm | |
70 sec. | |
Grade 91 |
Cellulose | |
10 μm | |
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. | |
205 μm |