missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes

Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.

Brand:  Cytiva 10401772


 View more versions of this product

Product Code. 11325324

  • £145.40 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.

  • Supports visual enumeration of bacteria
  • Appropriate for bacterial culturing, including E. coli culturing
  • Biologically inert with good thermal stability
  • Uniform microporous structure results in high flow rates when used for filtering
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
White
0.2 μm
20s/100mL/12.5cm2
Hydrophilic
ME24
135 μm
3.7bar
MCE
Sterile
Plain
Interleaving paper
100Pack
50 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes > Diameter: 50 mm, Sterile

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.