missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Small Roll White Wiper Dispenser

Suitable for use with the Kleenex™ Hand and Surface Sanitising Wipes to prevent the spread of bacteria in the workplace. Small Roll White Wiper Dispenser is ideal for high traffic environments such as offices, schools, gyms and restaurants.

Brand:  Kimberly-Clark Professional™ 7936

Product Code. 12601346

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Dispenses sheets of wiper roll without the need to touch the dispenser, helping to reduce contamination and the spread of germs
  • Quick-fill with a stylish white, easy-clean finish
  • Hassle-free maintenance
  • Ideal for high traffic environments such as offices, schools, gyms and restaurants
  • Can be wall mounted
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wiper Dispenser
15 x 15 x 29.3 cm
15 cm
15 cm
29.3 cm
Wall Mount
Hinged Lid with Front Opening
100 Pk.
White
5.9 in.
5.9 in.
11.5 in.
Plastic
1
Ideal for dispensing Kleenex hand and surface sanitizing wipes, in high traffic environments such as offices, schools, gyms and restaurants
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark Professional™ Small Roll White Wiper Dispenser > Dimensions (L x W x H): 29.3 x 15 x 15 cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.