Learn More
Cytiva Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes
Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.
Brand: Cytiva 10401706
90.16 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.
- Supports visual enumeration of bacteria
- Appropriate for bacterial culturing, including E. coli culturing
- Biologically inert with good thermal stability
- Uniform microporous structure results in high flow rates when used for filtering
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | |
Membrane Filter | |
0.2 μm | |
20s/100mL/12.5cm2 | |
Interleaving paper | |
100Pack | |
Filtration Membrane Disc |
3.7bar | |
MCE | |
Non-sterile | |
Plain | |
ME24 | |
135 μm | |
25 mm |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- ME24, 0.2µm 25mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
43Hours
6Minutes
59Seconds
21
Your input is important to us. Please complete this form to provide feedback related to the content on this product.