missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes
Facilitates the counting process and minimizes eye fatigue with color contrast between the surface and the colonies. Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes are characterized by a smooth and more uniform surface.
Brand: Cytiva 10401631
This item is not returnable.
View return policy
Description
- Excellent contrast for easier particle detection.
- Offers a high degree of internal surface area for greater adsorption of product.
- Higher dirt loading capacity.
- Biologically inert with good thermal stability.
- No surfactants to contaminate samples.
- Uniform microporous structure of membrane gives high flow rates.
Specifications
Autoclavable | |
Cellulose Acetate, Cellulose Nitrate | |
Non-sterile | |
Interleaving paper | |
25/Pk. | |
145 μm |
2.8 bar | |
0.45 μm | |
12.5 sec./100 mL/12.5 cm2 | |
ME25 | |
Plain | |
142 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ME25, 0.45µm 142mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction