missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Mixed Cellulose Ester Membranes: ME24

Recommended for the majority of routine applications

Brand:  Cytiva 10406972


 View more versions of this product

Product Code. 11355374

  • £146.67 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
  • Special nontoxic ink used is completely free from bacterial growth inhibitors
  • Manufactured under strictly controlled cleanroom conditions
  • Pore size: 0.2μm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Mixed Cellulose Ester
0.2 μm
To quantitatively evaluate microorganism counts
Black
Interleaving paper
0.31cm
266°F
Membrane
1.96 in.
White
ME 24
Sterile
ME 24
Hydrophilic
Medium
Grid
130°C
20 sec./100mL/12.5cm2
5 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers