Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Brand: Cytiva 7141-104
Description
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
Order Info
Autoclavable pack has 10 envelopes of 10 filters/pads for sterilization in the microbiological laboratory.

Specifications
Autoclavable | |
MCE | |
Sterile | |
Black | |
Individually Packed | |
100 Pack | |
Grid | |
130°C | |
47 mm |
White | |
0.45 μm | |
Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions | |
Pad, Interleaving paper | |
Medium | |
3.1mm | |
266°F | |
1.85 in. |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.