Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
£65.15 - £830.00
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Material (Membrane) | MCE |
Pore Size | 0.45 μm |
For Use With (Application) | Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions |
Product Code | Brand | Surface Type | Sterility | Includes | Packaging | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Type | Sterility | Includes | Packaging | Price | Quantity & Availability | ||
11914544
|
Cytiva
7141-154 |
Grid | Sterile | Pad, Interleaving paper | Individually Packed |
£830.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10153971
|
Cytiva
7141-004 |
Grid | Non-sterile | Pad | Pack |
£317.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11374824
|
Cytiva
7140-104 |
Plain | Sterile | Pad | Individually Packed |
£184.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11929207
|
Cytiva
7141-124 |
Grid | Sterile | Interleaving paper | Individually Packed |
£181.00
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10030461
|
Cytiva
7141-104 |
Grid | Sterile | Pad, Interleaving paper | Individually Packed |
£184.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10756262
|
Cytiva
7141-114 |
Grid | Sterile | Interleaving paper | Individually Packed |
£237.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11324834
|
Cytiva
7141-204 |
Grid | Non-sterile | Interleaving paper | Autoclave Pack |
£65.15
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
Order Info
Autoclavable pack has 10 envelopes of 10 filters/pads for sterilization in the microbiological laboratory.
Specifications
Autoclavable | |
MCE | |
Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions | |
3.1mm | |
130°C | |
47 mm |
White | |
0.45 μm | |
Medium | |
266°F | |
1.85 in. |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Your input is important to us. Please complete this form to provide feedback related to the content on this product.