Learn More
Cytiva Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Brand: Cytiva 7404-004
202.08 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 75
Additional Details : Weight : 0.03600kg
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing
Specifications
Autoclavable | |
Nylon | |
0.45 μm | |
100/Pk. | |
47 mm |
Membrane Filter | |
Nylon | |
Hydrophilic | |
Filtration Membrane Disc |