missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Brand: Cytiva 7404-009
228.71 GBP valid until 2025-12-31
Use promo code "24321" to get your promotional price.
This item is not returnable.
View return policy
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

Specifications
Autoclavable | |
0.45 μm | |
Hydrophilic | |
90 mm |
Nylon | |
For filtering aqueous solutions and most organic solvents | |
50/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Whatman™ Nylon Membranes > Pore Size: 0.45 μm, Diameter: 90 mm
Spot an opportunity for improvement?Share a Content Correction