missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Brand: Cytiva 7402-001
121.89 GBP valid until 2024-12-31
Use promo code "21931" to get your promotional price.
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing
Specifications
Autoclavable | |
0.2 μm | |
100/Pk. |
Nylon | |
Hydrophilic | |
13 mm |