missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nylon Membranes

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

Brand:  Cytiva 7404-001

130.68 GBP valid until 2025-12-31
Use promo code "24321" to get your promotional price.


 View more versions of this product

Product Code. 11394864

  • £144.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.

 

  • Pure nylon membranes
  • Tear-resistant
  • Filter many organic solvents
  • Autoclavable up to 135°C
  • Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
  • Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
0.5 μm
100/Pk.
Nylon
Hydrophilic
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Whatman™ Nylon Membranes > Pore Size: 0.45 μm, Diameter: 13 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.