missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap Disposable Capsules, 36 AS

Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.

Brand:  Cytiva 6706-3602

Code : 69

Additional Details : Weight : 0.13000kg

 View more versions of this product

Product Code. 11329094

  • £65.35 / Each
Estimated Shipment: 19-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
  • Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
  • Housing thermally fused (no glues, adhesives, or extraneous materials).
  • Integrity-testable by bubble point, pressure decay, or forward flow methods.
  • Provides highly effective filtration area in a small size.
  • Autoclavable; some presterilized with gamma irradiation.
  • Manufactured in clean room facilities under ISO Quality Systems.
Specifications

Specifications

Autoclavable
Nylon
Sterile
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
SB inlet
SB outlet plus filling bell
Polycap AS 36 Capsule Filter, sterile, 0.2 μm, SB inlet and outlet plus filling bell (1 pc)
0.2 μm
400 cm2
Polypropylene (PP)
4.1 bar (60 psi)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers