missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
Marke: Cytiva 2606T
Code : 69
Additional Details : Weight : 0.51000kg
Beschreibung
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.- First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
- Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
- Housing thermally fused (no glues, adhesives, or extraneous materials).
- Integrity-testable by bubble point, pressure decay, or forward flow methods.
- Provides highly effective filtration area in a small size.
- Autoclavable; some presterilized with gamma irradiation.
- Manufactured in clean room facilities under ISO Quality Systems.
Spezifikation
Polycap AS 36 Capsule Filter, 0.2 μm, FNPT inlet and outlet (5 pcs) | |
0.2 μm | |
400 cm2 | |
Polypropylene (PP) | |
4.1 bar (60 psi) |
Nylon | |
Non-sterile | |
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
FNPT | |
FNPT |