missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
Brand: Cytiva 6708-3602
This item is not returnable.
View return policy
Description
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.- First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
- Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
- Housing thermally fused (no glues, adhesives, or extraneous materials).
- Integrity-testable by bubble point, pressure decay, or forward flow methods.
- Provides highly effective filtration area in a small size.
- Autoclavable; some presterilized with gamma irradiation.
- Manufactured in clean room facilities under ISO Quality Systems.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | |
Nylon | |
Sterile | |
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
1/2 SB inlet | |
1/2 SB outlet |
Polycap AS 36 Capsule Filter, sterile, 0.2 μm, 1/2 SB inlet and outlet (1 pc) | |
0.2 μm | |
400 cm2 | |
Polypropylene | |
4.1 bar (60 psi) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Whatman™ Polycap Disposable Capsules, 36 AS > No.: 36 AS; Pore size; 0.2μm; Inlet/Outlet Connection: 3/8-1/2 in. hose barb
Spot an opportunity for improvement?Share a Content Correction