missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Offers high filtration efficiency and excellent filtrate purity
Brand: Cytiva 2810T
477.47 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Allows usage with broad range of solutions, pH, and temperature
- High flow and high retention capacity
- Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
- Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
Specifications
Polycap HD 150 Capsule Filter, 0.45 μm, with FNPT inlet and outlet (5 pcs) | |
Polycap HD 150 | |
With Female NPT inlet/outlet | |
5/Pk. |
0.45 μm | |
Polypropylene (PP) | |
4.1 bar (60 psi) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction