Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Offers high filtration efficiency and excellent filtrate purity
£425.00 - £577.92
Product Code | Brand | Pore Size | Inlet Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Inlet Style | Price | Quantity & Availability | ||||
11339104
|
Cytiva
2810T |
0.45 μm | With Female NPT inlet/outlet |
£577.92
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11399104
|
Cytiva
2812T |
5 μm | With Female NPT inlet/outlet |
£430.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11319114
|
Cytiva
2814T |
20 μm | With Female NPT inlet/outlet |
£430.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11359114
|
Cytiva
2813 |
10 μm | With 1/2 in. hose barb (HB) inlet/outlet |
£425.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11359104
|
Cytiva
2810 |
0.45 μm | With 1/2 in. hose barb (HB) inlet/outlet |
£425.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11339114
|
Cytiva
2813T |
10 μm | With Female NPT inlet/outlet |
£430.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Allows usage with broad range of solutions, pH, and temperature
- High flow and high retention capacity
- Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
- Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
Specifications
Polycap HD 150 | |
5/Pk. |