Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Offers high filtration efficiency and excellent filtrate purity
Brand:  Cytiva 2810
 View more versions of this product
Product Code. 11359104
  • £437.00 / Pack of 5
Explore more special offers
Description
Description
  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.45 μm
Polycap HD 150
With 1/2 in. hose barb (HB) inlet/outlet
5/Pk.