missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap™ TC High-Capacity Capsules

Developed for safe, efficient filtration of critical biological and tissue culture solutions. Whatman™ Polycap™ TC High-Capacity Capsules include PES membranes for excellent flow rate and protein absorption characteristics.

Brand:  Cytiva 6718-9582

154.00 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 11304634

  • £175.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Contain two layers of polyethersulfone membrane — inherently hydrophilic, low in extractables, excellent flowrates. Low protein-binding characteristics make capsules ideal for use with biologically sensitive solutions.

  • Pore size 0.8/0.2μm
  • Filtration area: 440, 930, or 1900cm2
  • Vent evacuates any air for fast filtration
  • Outlet is protected to prevent touch contamination during setup
  • Standard hose barb connections
  • Durable polypropylene filter housing can withstand maximum operating pressure of 4.1 bar (60psi)
  • Presterilized ISO-driven cleanroom manufacturing ensures product consistency
  • Stamped lot numbers ensure traceability of all critical components

Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.

Compliance

Pass HIMA Challenge Test for Sterilizing Grade Filters to retain >107CFU/cm2 Brevundimonas diminuta per ASTM™ 838-83 and FDA guidelines; microbial retention correlated to 100% integrity testing during manufacturing.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polycap TC 150 Capsule Filter, 0.8/0.2 μm, sterile, with 1/2 in SB inlet and outlet plus filling bell (1 pc + FB)
0.8/0.2 μm
1900 cm2
Polypropylene (PP)
4.1 bar (60 psi)
PES
Sterile
Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.
Stepped barb for 10 to 12mm tubing
Stepped barb for 6 to 10mm tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers