Learn More
Cytiva Whatman™ Polycap™ TC High-Capacity Capsules
Developed for safe, efficient filtration of critical biological and tissue culture solutions. Whatman™ Polycap™ TC High-Capacity Capsules include PES membranes for excellent flow rate and protein absorption characteristics.
£78.95 - £175.00
Specifications
Material (Membrane) | PES |
---|---|
Pore Size | 0.8/0.2 μm |
Sterility | Sterile |
For Use With (Application) | Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions. |
Housing Material | Polypropylene (PP) |
Product Code | Brand | Filtration Area | Inlet Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filtration Area | Inlet Style | Price | Quantity & Availability | ||||
11374624
|
Cytiva
6715-3682 |
440 cm2 | Stepped barb for 6 to 10mm tubing |
£78.95
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11384624
|
Cytiva
6715-7582 |
930 cm2 | Stepped barb for 6 to 10mm tubing |
£79.05
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11304634
|
Cytiva
6718-9582 |
1900 cm2 | Stepped barb for 10 to 12mm tubing |
£175.00
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Contain two layers of polyethersulfone membrane — inherently hydrophilic, low in extractables, excellent flowrates. Low protein-binding characteristics make capsules ideal for use with biologically sensitive solutions.
- Pore size 0.8/0.2μm
- Filtration area: 440, 930, or 1900cm2
- Vent evacuates any air for fast filtration
- Outlet is protected to prevent touch contamination during setup
- Standard hose barb connections
- Durable polypropylene filter housing can withstand maximum operating pressure of 4.1 bar (60psi)
- Presterilized ISO-driven cleanroom manufacturing ensures product consistency
- Stamped lot numbers ensure traceability of all critical components
Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.
Compliance
Pass HIMA Challenge Test for Sterilizing Grade Filters to retain >107CFU/cm2 Brevundimonas diminuta per ASTM™ 838-83 and FDA guidelines; microbial retention correlated to 100% integrity testing during manufacturing.
Specifications
PES | |
Sterile | |
Polypropylene (PP) | |
Stepped barb for 6 to 10mm tubing |
0.8/0.2 μm | |
Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions. | |
4.1 bar (60 psi) |