missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap™ TC High-Capacity Capsules

Developed for safe, efficient filtration of critical biological and tissue culture solutions. Whatman™ Polycap™ TC High-Capacity Capsules include PES membranes for excellent flow rate and protein absorption characteristics.

Brand:  Cytiva 6715-3682

Code : 69

Additional Details : Weight : 1.58760kg

 View more versions of this product

Product Code. 11374624

  • £78.95 / Each
Estimated Shipment: 07-08-2024
to see stock.

Description

Description

Contain two layers of polyethersulfone membrane — inherently hydrophilic, low in extractables, excellent flowrates. Low protein-binding characteristics make capsules ideal for use with biologically sensitive solutions.

  • Pore size 0.8/0.2μm
  • Filtration area: 440, 930, or 1900cm2
  • Vent evacuates any air for fast filtration
  • Outlet is protected to prevent touch contamination during setup
  • Standard hose barb connections
  • Durable polypropylene filter housing can withstand maximum operating pressure of 4.1 bar (60psi)
  • Presterilized ISO-driven cleanroom manufacturing ensures product consistency
  • Stamped lot numbers ensure traceability of all critical components

Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.

Specifications

Specifications

Polycap TC 36 Capsule Filter, 0.8/0.2 μm, sterile, with SB inlet and outlet plus filling bell (1 pc + FB)
0.8/0.2 μm
440 cm2
Polypropylene (PP)
4.1 bar (60 psi)
PES
Sterile
Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.
Stepped barb for 6 to 10mm tubing
Stepped barb for 6 to 10mm tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers