Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran
100% pure nitrocellulose to ensure the highest binding capacity possible
Brand:  Cytiva 10402452
 View more versions of this product
Product Code. 11395324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
  • Also available in circular formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 cm
20 x 20cm
High
Sheet
0.2 μm
20 cm
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
5/Pk.
Nitrocellulose