missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles

100% pure nitrocellulose to ensure the highest binding capacity possible

Brand:  Cytiva 10402525

320.59 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.



 View more versions of this product

Product Code. 11375334

  • £411.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Universal membrane suitable for Western, Southern, and Northern blotting
  • Hydrophilic, so no prewetting with methanol is required
  • High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
  • Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
  • Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
  • Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
132 mm
Disc
0.45 μm
High
50/Pk.
Nitrocellulose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger||||Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.