missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Puradisc 25 Syringe Filters

Ideal for quick and efficient filtration of samples

Brand:  Cytiva 6757-2502

Additional Details : Weight : 0.58000kg

 View more versions of this product

Product Code. 16675695

  • £362.00 / Pack of 200
Estimated Shipment: 09-10-2024
to see stock.
Add to basket

Description

Description

  • Pigment-free polypropylene
  • Standard inlet and outlet luer connectors
  • Tube-tip format (optional) for accurate dispensing into a micro-vial
  • Wide range of membranes
  • Sterile option for critical applications
Specifications

Specifications

Regenerated Cellulose
Filtration
200/Pk.
Syringe Filter
0.2 μm
Polypropylene
Whatman™
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers