missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

Brand:  Cytiva 6790-2502

1284.00 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.



Code : A0

Additional Details : Weight : 2.54000kg

 View more versions of this product

Product Code. 11373305

  • £1375.00 / Pack of 1000
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121ºC
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications

Specifications

Dimensions: 22.9 × 28.4mm
0.2 μm
4.2 cm2
Female Luer lock
Male Luer
Up to 100 mL
Polypropylene
Non-sterile
Polypropylene
1000
Puradisc Syringeless Filter
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers