missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
£144.00 - £1375.00
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 4.2 cm2 |
Volume (Metric) | Up to 100 mL |
Diameter (Metric) | 25 mm |
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
11304694
|
Cytiva
6786-2502 |
0.2 μm | 50 |
£144.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11373305
|
Cytiva
6790-2502 |
0.2 μm | 1000 |
£1375.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11313315
|
Cytiva
6790-2504 |
0.45 μm | 100 |
£1110.00
Pack of 1000 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11354694
|
Cytiva
6788-2504 |
0.45 μm | 200 |
£508.78
Pack of 200 |
In Stock
Log in to see stock. |
||||
11344694
|
Cytiva
6788-2502 |
0.2 μm | 200 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11314694
|
Cytiva
6786-2504 |
0.45 μm | 50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
- Diameter: 25mm
- Sample volume up to 100mL
- Low hold-up volumes for maximum sample recovery
- Optional glass microfiber
- Autoclave at 121ºC
Applications:
- HPLC aqueous sample preparation
- Biological sample preparation
- Buffer solutions
- Salt solutions
- Tissue culture media
- Irrigation solutions
- Sterile isolation
Specifications
Non-sterile | |
4.2 cm2 | |
Up to 100 mL | |
25 mm |