Learn More
Cytiva Whatman™ Quartz Air Sampling Filters, Grade QM-A, Circle
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
£217.00 - £829.00
Product Code | Brand | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Price | Quantity & Availability | |||||
11330624
|
Cytiva
1851-025 |
25 mm |
£237.00
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
11303345
|
Cytiva
1851-101 |
101.6 mm |
£474.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11313355
|
Cytiva
1851-085 |
85 mm |
£466.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11393345
|
Cytiva
1851-050 |
50 mm |
£293.09
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11343345
|
Cytiva
1851-082 |
82 mm |
£365.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11383345
|
Cytiva
1851-150 |
150 mm |
£829.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11353345
|
Cytiva
1851-090 |
90 mm |
£525.00
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
11393335
|
Cytiva
1851-045 |
45 mm |
£285.86
Pack of 100 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11373345
|
Cytiva
1851-118 |
118 mm |
£610.00
Pack of 100 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
11313345
|
Cytiva
1851-032 |
32 mm |
£217.00
Pack of 100 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
11340624
|
Cytiva
1851-037 |
37 mm |
£266.00
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
10455211
|
Cytiva
1851-047 |
47 mm |
£381.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11323345
|
Cytiva
1851-055 |
55 mm |
£296.00
Pack of 100 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Description
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
- Temperature resistant up to 500°C
- Binder-free
- High loading capacity
- Fast flow rates (air flow rate 6.4 s/100 ml/in2)
- Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols
- Filters are suitable for PM-10 sampling and analysis
Specifications
Circle | |
Grade QM-A |