missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ TurboBlotter Transfer System, Large Kits

Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA

Brand:  Cytiva 10416324

Code : 69

Additional Details : Weight : 4.38000kg

 View more versions of this product

Product Code. 11909307

  • £492.00 / Each
Estimated Shipment: 16-09-2024
to see stock.
Add to basket

Description

Description

  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable

  • Model is 21 × 26cm, for gels from 12 × 21cm to 20 × 25cm

Order Info

Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).

Specifications

Specifications

20 x 25 cm Gel TurboBlotter™ Transfer System
Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
25 cm
20 cm
Nytran™ SPC 0.45μm TurboBlotter™ Kit
9.8 in.
7.8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers