missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ TurboBlotter Transfer System, Large Kits
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
£348.00 - £492.00
Specifications
Type | Nytran™ SPC 0.45μm TurboBlotter™ Kit |
---|---|
Includes | Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials. |
Includes: Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytrantrademark (TM) SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Product Code | Brand | Length (English) Gel | Length (Metric) Gel | Width (English) Gel | Width (Metric) Gel | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) Gel | Length (Metric) Gel | Width (English) Gel | Width (Metric) Gel | Price | Quantity & Availability | ||
11969297
|
Cytiva
10416316 |
7.8 in. | 20 cm | 5.9 in. | 15 cm |
£348.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
11909307
|
Cytiva
10416324 |
9.8 in. | 25 cm | 7.8 in. | 20 cm |
£492.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
Description
- Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
- Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
- Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
- Very easy to set up and works without power or vacuum source
- Requires less buffer and blotting paper than conventional transfers
- Reusable
- Model is 21 × 26cm, for gels from 12 × 21cm to 20 × 25cm
Order Info
Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).
Specifications
Nytran™ SPC 0.45μm TurboBlotter™ Kit | |
Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials. |