Learn More
Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL
Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis
Brand: Cytiva 7700-9901
895.36 GBP valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Code : 69
Additional Details : Weight : 2.72160kg
Description
- Natural polypropylene plates
- Patented integral filter design prevents well-to-well cross contamination
- Standard footprint—compatible with most plate readers, autosamplers and robots
- Unique drip director design on the bottom of the plates controls filtrate placement
- Available in several media types
Specifications
Standard | |
4 x 6 | |
Natural | |
24 | |
Filter | |
Long drip director and Glass Microfiber GF/C Filter |
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation. | |
1.2 μm | |
Polypropylene | |
Non-sterile | |
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation |
Safety and Handling
- FILTER GLASS CIRCLE
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : glass, oxide, chemicals
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.