missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL
Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis
£795.00 - £987.00
Specifications
Format | Standard |
---|---|
Description | The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation. |
Array | 4 x 6 |
Material | Polypropylene |
No. of Wells | 24 |
Product Code | Brand | Color | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Pore Size | Price | Quantity & Availability | ||||
11965296
|
Cytiva
7700-9904 |
Clear | 25 to 30 μm |
£795.00
Pack of 25 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
11919134
|
Cytiva
7700-9917 |
Natural | 10 to 12 μm |
£795.00
Pack of 25 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
11328674
|
Cytiva
7700-9901 |
Natural | 1.2 μm |
£987.00
Pack of 25 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
Description
- Natural polypropylene plates
- Patented integral filter design prevents well-to-well cross contamination
- Standard footprint—compatible with most plate readers, autosamplers and robots
- Unique drip director design on the bottom of the plates controls filtrate placement
- Available in several media types
Specifications
Standard | |
4 x 6 | |
24 | |
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation |
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation. | |
Polypropylene | |
Filter |