missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL
Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis
Marca: Cytiva 7700-9904
Code : A0
Dettagli aggiuntivi : Weight : 2.61000kg
Descrizione
- Natural polypropylene plates
- Patented integral filter design prevents well-to-well cross contamination
- Standard footprint—compatible with most plate readers, autosamplers and robots
- Unique drip director design on the bottom of the plates controls filtrate placement
- Available in several media types
Specifica
Standard | |
4 x 6 | |
Clear | |
Polypropylene | |
24 | |
Filter | |
Long drip director and MBPP Filter |
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation. | |
25 to 30 μm | |
UNIFILTER Microplate, 24-well, 10 mL, 25 to 30 μm melt blown polypropylene, natural polypropylene | |
10,000 μL | |
Non-sterile | |
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation | |
25/Pk. |