missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL

Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis

Brand:  Cytiva 7700-9917

Code : A0

Additional Details : Weight : 2.84000kg

 View more versions of this product

Product Code. 11919134

  • £795.00 / Pack of 25
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Natural polypropylene plates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types

Specifications

Specifications

Standard
4 x 6
Natural
Polypropylene
24
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
10 to 12 μm
UNIFILTER Microplate, 24-well, 10 mL, 10 to 12 μm melt blown polypropylene, natural polypropylene
10,000 μL
Filter
Long drip director and MBPP Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers