missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 60mL BOD Bottles with Glass Pennyhead Stoppers
Securely store samples with these BOD bottles, which include pennyhead-style stoppers
£1165.00 - £1520.00
Specifications
Volume (Metric) | 60 mL |
---|---|
For Use With (Application) | Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action |
Marking Spot | Yes |
Material | Borosilicate Glass |
Stopper | Pennyhead |
Product Code | Brand | No. Sequence | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. Sequence | Price | Quantity & Availability | |||||
12413711
|
DWK Life Sciences
227494-00G |
Unnumbered |
£1165.00
Pack of 36 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12483711
|
DWK Life Sciences
227494-99G |
Specials |
£1520.00
Pack of 36 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12423711
|
DWK Life Sciences
227494-01G |
1 to 36 |
£1375.00
Pack of 36 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12443711
|
DWK Life Sciences
227494-02G |
37 to 72 |
£1375.00
Pack of 36 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
12463711
|
DWK Life Sciences
227494-03G |
73 to 108 |
£1375.00
Pack of 36 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
- Type I borosilicate glass construction
- Height: 115mm (4.53 in.)
- Diameter: 43mm (1.69 in.)
- Capacity: 60mL (2 oz.)
Specifications
60 mL | |
Yes | |
Pennyhead | |
4.5 in. | |
1.69 in. |
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action | |
Borosilicate Glass | |
2.02 oz. | |
11.5 cm | |
4.3 cm |