Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
GREENER_CHOICE
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand:  DWK Life Sciences 224193-05
74.01 GBP valid until 2024-12-31
Use promo code "21972" to get your promotional price.
 View more versions of this product
Product Code. 10326902
  • £123.00 / Pack of 1000
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
1000/Cs.
20 mm
Aluminum
Complete Tear-off Cap