missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Borosilicate Glass Coliwasa Samplers

Offer excellent chemical resistance and can be cleaned repeatedly

Brand:  DWK Life Sciences 885231

 View more versions of this product

Product Code. 12497301

  • £575.00 / Pack of 12
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PTFE seal permits sampling to within 0.5 in. (1.3cm) of drum bottom
  • Positive seal allows transfer of sample to bottle without any loss
  • A 200mL sample may be obtained from a full 55 gal. (208L) drum
  • Available prescored for disposal after a single use or unscored for repeated use

Order Info

Capacity: 6.8 oz. (200mL); Dimensions: 0.88 in. dia. x 42 in.L (2.2 x 107cm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prescored Coliwasa Sampler
PTFE
200 mL
0.86 in.
2.2 cm
106.6 cm
Borosilicate Glass
6.76 oz.
0.5 in.
1.3 cm
42 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.