Learn More
DWK Life Sciences Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures
Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.
£30.15 - £493.00
Specifications
Color Amber
Material Glass
Closure Color White
Closure Liner Poly/Vinyl
Closure Material Polypropylene
View More Specs
Products 12
Product Code Brand Capacity (Metric) Closure Size Quantity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Quantity Price Quantity & Availability  
12349859
View Documents
DWK Life Sciences
W216954
30 mL 28-400 mm 432/Cs.
£493.00
Pack of 432
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
11784189
View Documents
DWK Life Sciences
W216939
30 mL 28-400 mm 24/Cs.
£30.15
Pack of 24
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
12359859
View Documents
DWK Life Sciences
W216955
60 mL 33-400 mm 216/Cs.
£290.00
Pack of 216
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
11774189
View Documents Promotion Details
DWK Life Sciences
W216940
60 mL 33-400 mm 24/Cs.
£35.46
Pack of 24
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
12369859
View Documents
DWK Life Sciences
W216956
125 mL 38-400 mm 180/Cs.
£298.00
Pack of 180
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
10689895
View Documents Promotion Details
DWK Life Sciences
125 mL 38-400 mm 24/Cs.
£39.50
Pack of 24
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
11714199
View Documents
DWK Life Sciences
W216942
250 mL 45-400 mm 24/Cs.
£48.35
Pack of 24
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
12379859
View Documents
DWK Life Sciences
W216957
250 mL 45-400 mm 84/Cs.
£175.00
Pack of 84
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
12389859
View Documents
DWK Life Sciences
W216958
500 mL 53-400 mm 60/Cs.
£163.00
Pack of 60
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
11714219
View Documents
DWK Life Sciences
W216943
500 mL 53-400 mm 12/Cs.
£43.70
Pack of 12
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
11759665
View Documents
DWK Life Sciences
W216944
1000 mL 53-400 mm 12/Cs.
£186.00
Pack of 12
Estimated Shipment: 02-07-2025
to see stock.
Add to basket
12399859
View Documents
DWK Life Sciences
W216959
1000 mL 53-400 mm 36/Cs.
£136.00
Pack of 36
Estimated Shipment: 02-10-2025
to see stock.
Add to basket
Description
Description
Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.
  • USP Type III soda-lime glass
  • Allows for easy removal of contents
Specifications
Specifications
Amber
White
Polypropylene
Glass
Poly/Vinyl
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures