Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand:  DWK Life Sciences W988008-S
535.29 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17952621
  • £580.00 / Pack of 125
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
OmniFlex 3G Igloo
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
125/Cs.
Wheaton CompletePAK
20 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers > 20 mm OmniFlex 3G Igloo Lyo Stopper, 125/Cs.