missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Deck Kit for Compact Roller System for Small Bottles

Each deck holds two roller bottles

Brand:  DWK Life Sciences 348930

 View more versions of this product

Product Code. 10579311

  • £671.57 / Each
Estimated Shipment: 14-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Attaches quickly and easily to Wheaton Compact Roller System for Small Bottles (above), which will accept two additional decks
  • Kit includes assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions

Order Info

Weight: 5 lb. (2.3kg)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Deck Kit
4 kg
Compact small bottle roller culture apparatus
9 lb.
Charcoal
Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Deck Kit for Compact Roller System for Small Bottles > Deck Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.