Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
Brand:  DWK Life Sciences 224814
 View more versions of this product
Product Code. 13171803
  • £176.00 / Pack of 200
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Order Info
    Caps available separately
    Compliance
    Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Amber
    Borosilicate Glass
    Screw
    Autoclavable
    60 mm
    Sample Vial in Lab File
    17 mm
    8 mL
    2.2 dr.
    Screw Cap
    USP Type I
    M-T File™ and Vial Rack
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products