missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Hybridization Bottles
For use in nucleic acid and protein blotting procedures that require a minimum of probe solution
Brand: DWK Life Sciences 805011
This item is not returnable.
View return policy
Description
- Manufactured from heavy-wall borosilicate glass
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
- Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
Order Info
Items come as one per case
Specifications
USP Type I | |
45 mm | |
2.75 in. | |
Nucleic acid and protein blotting procedures that require a minimum of probe solution | |
PTFE or Silicone | |
300 mm | |
Hybridization Bottle |
Polycarbonate | |
Plastisol | |
70 mm | |
11.81 in. | |
Screw | |
Borosilicate Glass |