missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Hybridization Bottles
For use in nucleic acid and protein blotting procedures that require a minimum of probe solution
£70.80 - £142.00
Specifications
Certifications/Compliance | USP Type I |
---|---|
Closure Material | Polycarbonate |
Closure Size | 45 mm |
Coating | Plastisol |
Closure Type | Screw |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Product Type | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Product Type | Material | Price | Quantity & Availability | ||
11541325
|
DWK Life Sciences
805003 |
35 mm | 150 mm | Hybridization Bottle | Borosilicate Glass |
£79.60
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
11724209
|
DWK Life Sciences
805000 |
35 mm | 75 mm | Hybridization Bottle | Borosilicate Glass |
£70.80
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
10717893
|
DWK Life Sciences
805007 |
35 mm | 300 mm | Hybridization Bottle | Borosilicate Glass |
£83.95
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
12437181
|
DWK Life Sciences
805027 |
35 mm | 300 mm | Safety Coated Hybridization Bottle | Borosilicate Glass |
£106.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
12417181
|
DWK Life Sciences
805020 |
35 mm | 75 mm | Safety Coated Hybridization Bottle | Borosilicate Glass |
£93.25
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
11531325
|
DWK Life Sciences
805001 |
35 mm | 100 mm | Hybridization Bottle | Borosilicate Glass |
£77.10
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
12871474
|
DWK Life Sciences
805021 |
35 mm | 100 mm | Safety Coated Hybridization Bottle | Borosilicate Glass |
£97.60
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
11551325
|
DWK Life Sciences
805023 |
35 mm | 150 mm | Safety Coated Hybridization Bottle | Borosilicate Glass |
£101.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
12447181
|
DWK Life Sciences
805031 |
70 mm | 300 mm | Safety Coated Hybridization Bottle | Borosilicate Glass |
£142.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
12742447
|
DWK Life Sciences
805011 |
70 mm | 300 mm | Hybridization Bottle | Borosilicate Glass |
£107.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||
Description
- Manufactured from heavy-wall borosilicate glass
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
Order Info
Items come as one per case
Specifications
USP Type I | |
45 mm | |
Screw | |
Nucleic acid and protein blotting procedures that require a minimum of probe solution |
Polycarbonate | |
Plastisol | |
Borosilicate Glass | |
PTFE or Silicone |