missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Liquid Scintillation Vials

Provides an economical alternative to standard scintillation vials. Wheaton™ Liquid Scintillation Vials are manufactured from rigid yet flexible HDPE plastic.

Brand:  DWK Life Sciences 986706

 View more versions of this product

Product Code. 12477341

  • £339.00 / Pack of 500
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provides an economical alternative to standard scintillation vials. Wheaton™ Liquid Scintillation Vials are manufactured from rigid yet flexible HDPE plastic.

  • With urea caps
  • With metal foil/cork liner

Order Info

O.D. x H: 1.125 x 2.375 in. (27 x 61mm) with cap. Minimum neck I.D., 0.5 in. (12.3mm). Packaged in utility trays with each partitioned tray holding 100 vials. Packed with caps attached or caps packaged separately. Cases of 1000 vials are packed in a single plastic bag with caps in a separate bag.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyethylene
1.06 in.
White
HDPE
2.4 in.
Tray
Urea
61 mm
0.5 in.
12.7 mm
27 mm
20 mL
500
0.67 oz.
Attached Screw Cap
500/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers