Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Brand:  DWK Life Sciences W228790
148.69 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15880130
  • £206.96 / Pack of 6
Explore more special offers
Description
Description
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hinged Lid
Plastic
Convenient Storage and Transportation of Sample Vials
54 (2, 4 and 8 mL Vials)
54
6 Case
Clear
Rectangular
54 x 8mL vials
Foam Insert, Index Card, Partition, Polystyrene Case
Storage Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories > Vial file; Holds up to fifty four 8mL vials