Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
Brand:  DWK Life Sciences 356916
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 12401431
  • £196.00 / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass flasks
  • Two large side arms with screw-cap closures
  • Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
  • Graduated
  • Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
  • Autoclavable flasks and impellers for selected sizes can also be ordered separately
  • U.S. Patent No. 3,854,704
Specifications
Specifications
125 mL
Flask only
Autoclavable
51-440
Borosilicate Glass
33-430
Yes