Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
£196.00 - £1195.00
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
10110631
|
DWK Life Sciences
356834 |
1000 mL | Flask with impeller |
£690.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10488512
|
DWK Life Sciences
356832 |
500 mL | Flask with impeller |
£475.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10202441
|
DWK Life Sciences
356830 |
125 mL | Flask with impeller |
£317.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10469862
|
DWK Life Sciences
356831 |
250 mL | Flask with impeller |
£449.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
12401431
|
DWK Life Sciences
356916 |
125 mL | Flask only |
£196.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
12421431
|
DWK Life Sciences
356922 |
500 mL | Flask only |
£396.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10684891
|
DWK Life Sciences
356839 |
6 L | Flask with impeller |
£1195.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10789341
|
DWK Life Sciences
356837 |
3000 mL | Flask with impeller |
£885.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
Description
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifications
Yes |