Learn More
DWK Life Sciences Wheaton™ Omnispense™ Plus Peristaltic Pump Accessory Pump Heads
For use with Wheaton Omnispense Plus dispensing pump
Brand:  DWK Life Sciences W375025
Code : 03
Additional Details : Weight : 0.68000kg
 View more versions of this product
Product Code. 12472132
  • £1150.00 / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
  • Stackable pump heads (13-687-444 and -442) allow user to add up to two additional pumpheads, for a total of three pump heads per dispensing pump
  • Cassette pump head ideal for multichannel applications
  • Removable cassettes can be preloaded with tubing elements and changed without stopping the drive
†For tubing with bore sizes from 0.13 to 2.79mm.
Specifications
Specifications
Optional 5-Channel Pump Head for use with Small Bore Tubing
4
Cassette Pump Head
Omnispense™ plus peristaltic pump
5