Learn More
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle.
£95.30 - £211.00
Specifications
Material Silicone
Thickness (English) 0.09 in.
Thickness (Metric) 2.3 mm
View More Specs
Products 8
Product Code Brand Diameter (Metric) Inner Overall Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Overall Length (Metric) Price Quantity & Availability  
12411531
View Documents
DWK Life Sciences
374308
8 mm 760 cm
£197.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
10118471
View Documents
DWK Life Sciences
374314
8 mm 150 cm
£197.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
10352392
View Documents
DWK Life Sciences
374313
6 mm 150 cm
£190.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
12827913
View Documents
DWK Life Sciences
374311
3 mm 150 cm
£148.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
11714269
View Documents
DWK Life Sciences
374304
2 mm 760 cm
£154.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
10256862
View Documents
DWK Life Sciences
374310
2 mm 150 cm
£95.30
Each
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
10107624
View Documents
DWK Life Sciences
374305
3 mm 760 cm
£165.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
12401531
View Documents
DWK Life Sciences
374306
6 mm 760 cm
£211.00
Each
Estimated Shipment: 25-07-2025
to see stock.
Add to basket
Description
Description
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle
Specifications
Specifications
Silicone
0.09 in.
2.3 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use