missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Prescored Clear Standards Ampules

Glass containers designed to be hermetically sealed, preserving purity and extending content shelf life

Brand:  DWK Life Sciences 176759

 View more versions of this product

Product Code. 11748709

  • £398.00 / Pack of 25
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

A gold band on the stem indicates that the ampule is pre-scored to facilitate snapping.

  • Constructed of USP Type I clear and amber low-extractable borosilicate glass
  • Methods of sterilization: autoclave, dry heat and EtO
  • Prevents cross contamination

Standards packaging, environmental and diagnostic standards, lyophilization and cryogenic storage

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7 mL
Prescored Ampule
1.7 cm
9.5 cm
ASTM E438 Type I, Class A
Borosilicate Glass
Clear
0.66 in.
3.74 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.